Learn More
CTL Scientific Supply Corp Reagent VISOCOLOR Powder Pillows for photometric determination of free Chlorine from 0.03 - 6.00 mg/L. , contains 1,000 Powder Pillows
SDP
Supplier:  CTL Scientific Supply Corp 936220.1
Encompass_Preferred
Reagent powder pillows for photometric determination of free chlorine from 0.03 - 6.00 mg/L. Powder pillows combine easy dosing with photometric precision. Ideal for pool or drinking water analysis.
Catalog No. NC2958032