Learn More
CTL Scientific Supply Corp VISOCOLOR ECO FREE CHLORINE 2
SDP
Supplier:  CTL Scientific Supply Corp 931016
Encompass_Preferred
Environmental test kit for free Chlorine Gradation of scale <0.1-0.1-0.2-0.3-0.4-0.6-0.9-1.2-2.0 m/L 150 tests
Catalog No. 50-300-366