Learn More
CTL Scientific Supply Corp VISOCOLOR ECO MAGANESE
SDP
Supplier:  CTL Scientific Supply Corp 931038
Encompass_Preferred
Environmental test kit for Maganese Gradation of scale: 0.1 - 0.2 - 0.3 - 0.5 - 0.7 - 0.9 - 1.2 - 1.5 mg/l 170 tests/kit
Catalog No. 50-300-377