Learn More
CTL Scientific Supply Corp VISOCOLOR ECO SILICA
SDP
Supplier:  CTL Scientific Supply Corp 931033
Encompass_Preferred
Environmental test kit for Silica Gradation of scale: 0.2 - 0.4 - 0.6 - 1.0 - 1.5 - 2.0 - 2.5 - 3.0 mg/l 100 tests/kit
Catalog No. 50-300-375