Learn More
Selleck Chemical LLC Visomitin SKQ1-10mg
SDP
Catalog No. 502389874
Encompass_Preferred
Visomitin (SKQ1) is a novel mitochondrial-targeted antioxidant that holds promise in the treatment of inflammation associated with ocular surface diseases such as dry eye disease (DED) and corneal wounds. SkQ1 shows a TC50 of 317 nM on HCjE cells.
Catalog No. 50-238-9874 Supplier Selleck Chemical LLC Supplier No. S972910MG
Visomitin (SKQ1) is a novel mitochondrial-targeted antioxidant that holds promise in the treatment of inflammation associated with ocular surface diseases such as dry eye disease (DED) and corneal wounds. SkQ1 shows a TC50 of 317 nM on HCjE cells.