Learn More
Med Vet International Visor Light Replacement Bulb, J0801VD1
Catalog No. 5000583971
Encompass_Preferred
OptiVisor is a precision binocular headband magnifier which permits unrestricted user efficiency— leaving both hands free, while reducing eye strain—allowing three dimensional vision. It can be worn over prescription eyeglasses. Equipped with these unique features:• Optical glass prismatic lenses. Ground and polished to precision optical standards. Bevel edged and mounted in interchangeable plastic frame. with Adjustable Headband.• Adjustable pivots allow the visor to be tilted out of the way when not in use, and at your fingertips when magnification is needed.• Adjustable headband conforms to fit all head sizes. Fabricated from a special blend of lightweight material. Comfortable when worn for long periods of time.• High impact visor is tough, durable, and always new in appearance.• Genuine leather comfort band is padded with orthopedic felt to add comfort.• Magnifies 2.5 times at 8”.• Optivisor is perfect for eye exams, (i.e. corneal foreign body, distichiasis).
Catalog No. 50-005-83971 Supplier Med Vet International Supplier No. JOR801VD1
OptiVisor is a precision binocular headband magnifier which permits unrestricted user efficiency— leaving both hands free, while reducing eye strain—allowing three dimensional vision. It can be worn over prescription eyeglasses. Equipped with these unique features:• Optical glass prismatic lenses. Ground and polished to precision optical standards. Bevel edged and mounted in interchangeable plastic frame. with Adjustable Headband.• Adjustable pivots allow the visor to be tilted out of the way when not in use, and at your fingertips when magnification is needed.• Adjustable headband conforms to fit all head sizes. Fabricated from a special blend of lightweight material. Comfortable when worn for long periods of time.• High impact visor is tough, durable, and always new in appearance.• Genuine leather comfort band is padded with orthopedic felt to add comfort.• Magnifies 2.5 times at 8”.• Optivisor is perfect for eye exams, (i.e. corneal foreign body, distichiasis).