Learn More
Grainger Ergodyne Chill-Its Hard Hat Brim With Neck Shade, Polyester, Orange, 1 ea
Catalog No. 502795596
Encompass_Preferred
This hard hat brim and neck shade provides full coverage to protect the face and back of the neck from sun damage. Constructed from 100% lightweight polyester, it attaches securely to most hard hats to keep workers comfortable in environments lacking shade.
  • Provides high visibility in orange to help keep workers safe and seen
  • Features a durable, plastic brim that holds its shape
  • Offers universal fit with an elastic inner rim and hook and loop attachment straps
  • Constructed from 100% polyester for durability and comfort
  • Designed for protection against sun and UV rays to help mitigate heat stress and sunburn
  • Hand washable for easy care
  • Brim dimensions: 8.3 cm (3.25 in.) L
  • Shade dimensions: 24.8 cm (9.75 in.) L x 49.6 cm (19.5 in.) W
Catalog No. 50-279-5596 Supplier Grainger Supplier No. 19TZ64
This hard hat brim and neck shade provides full coverage to protect the face and back of the neck from sun damage. Constructed from 100% lightweight polyester, it attaches securely to most hard hats to keep workers comfortable in environments lacking shade.
  • Provides high visibility in orange to help keep workers safe and seen
  • Features a durable, plastic brim that holds its shape
  • Offers universal fit with an elastic inner rim and hook and loop attachment straps
  • Constructed from 100% polyester for durability and comfort
  • Designed for protection against sun and UV rays to help mitigate heat stress and sunburn
  • Hand washable for easy care
  • Brim dimensions: 8.3 cm (3.25 in.) L
  • Shade dimensions: 24.8 cm (9.75 in.) L x 49.6 cm (19.5 in.) W