Learn More
Andwin Scientific VISORGOG FULL VIS SCR VENT BLU
SDP
Catalog No. NC9668414
Encompass_Preferred
The Visorgogs Safety Goggles provide superior protection for eyes against flying particles or splashing liquids. This product features a comfortable fit and a vented, opaque blue visor in order to shade eyes and lessen eye strain and fatigue. The lenses are constructed to have a down-angle vision for clear visibility. These goggles may be worn over eyeglasses.
This item is not returnable. View return policy
Catalog No. NC9668414 Supplier Andwin Scientific Supplier No. 33012006
The Visorgogs Safety Goggles provide superior protection for eyes against flying particles or splashing liquids. This product features a comfortable fit and a vented, opaque blue visor in order to shade eyes and lessen eye strain and fatigue. The lenses are constructed to have a down-angle vision for clear visibility. These goggles may be worn over eyeglasses.