Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leica Microsystems Visoria B Laboratory Microscope for Immunofluorescence

Catalog No. 14823449 Shop All Leica Microsystems Products
Change view
Click to view available options
Magnification Power:
5X to 100X
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
14-823-449 5X to 100X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-823-449 Supplier Leica Microsystems Supplier No. 111000017
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This microscope is suitable for applications in life sciences and clinical laboratories for immunofluorescence studies. Workflows can be streamlined with encoded functions, optimized light settings, and other features.

  • Visoria B Laboratory Microscope (Research use only) with illumination management
  • Trinocular phototube
  • Two eyepieces (10x), field of view 22 mm
  • K3M microscope camera, includes c-mount 0.5x
  • High-power white LED and 4-channel LED light source
  • Fluorescence axis, 5-position, with four filters (Y5, I3, N2.1, DAPI )
  • Contrast methods: Brightfield, Fluorescence
  • Encoded nosepiece, 6x
  • Objectives (N PLAN): 5x, 10x, 20x, 40x, 50x, 100x
  • ErgoStage with ceramic plate, with specimen holder
  • 2-step focus drive with height-adjustable focus knobs
  • Condenser A 0.9 (P)
  • Immersion liquid, ISO 8036
  • Dust protection cover
  • Quick start guide and user manual
  • Efficiency through encoded functions
  • More comfort with ergonomics
  • Visualize sample details with the right contrast
  • Powered by Enersight software platform
TRUSTED_SUSTAINABILITY

Specifications

Eyepiece 10X/22 mm
Head Style Trinocular, 30° Inclined
Lighting Type LED
Magnification Power 5X to 100X
Model Visoria B Fluo
Objective Types Plan
Product Type Trinocular Fluorescent Microscope
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.