Learn More
AdipoGen VISTA [B7-H5] (human) (rec.) (His) (10 µg)
SDP
Supplier:  AdipoGen AG40B0177C010
Encompass_Preferred
Protein. The extracellular domain of human VISTA (B7-H5) (aa 33-194) is fused at the C-terminus to a His-tag. Source: HEK 293 cells. Endotoxin: <0.01EU/µg. Lyophilized. Contains PBS. Human VISTA is a 55 to 65kDa type I Ig membrane protein with the extracellular domain homologous to PD-L1. VISTA is mainly expressed on hematopoietic tissues (spleen, thymus and bone marrow) and on myeloid cells with lower expression on CD4+ and CD8+ T cells. VISTA is a new negative checkpoint regulator that potently suppresses T cell activation. Overexpression of VISTA on tumors inhibits the protective anti-tumor immunity and blockade of VISTA enhances antitumor immunity in multiple tumor models. VISTA on antigen presenting cells (APC) delivers negative signaling to T cells via a yet-to-identify binding partner on T cells.
Catalog No. 50-205-3278