Learn More
Medchemexpress LLC VISTA/B7-H5 Mouse 100ug
SDP
Catalog No. 5000180129
Encompass_Preferred
VISTA/B7-H5 Protein is a type I transmembrane protein expressed primarily in white blood cells that inhibits T cell function VISTA/B7-H5 Protein promotes embryonic stem cell differentiation by inhibiting BMP4 signaling and stimulates MMP14 mediated MMP2 activation VISTA/B7-H5 Protein is highly expressed in tumors VISTA/B7-H5 Protein Mouse (HEK293 Fc) is the recombinant mouse-derived VISTA/B7-H5 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-80129 Supplier Medchemexpress LLC Supplier No. HYP73547100UG
Add to Cart
Add to Cart
VISTA/B7-H5 Protein is a type I transmembrane protein expressed primarily in white blood cells that inhibits T cell function VISTA/B7-H5 Protein promotes embryonic stem cell differentiation by inhibiting BMP4 signaling and stimulates MMP14 mediated MMP2 activation VISTA/B7-H5 Protein is highly expressed in tumors VISTA/B7-H5 Protein Mouse (HEK293 Fc) is the recombinant mouse-derived VISTA/B7-H5 protein expressed by HEK293 with C-hFc labeled tag