Learn More
Medchemexpress LLC VISTA/B7-H5 Rat HE 10ug
SDP
Catalog No. 5000180141
Encompass_Preferred
VISTA/B7-H5 protein an immunoregulatory receptor inhibits T-cell response and may regulate embryonic stem cell differentiation by inhibiting BMP4 signaling It also stimulates MMP14-mediated MMP2 activation suggesting a role in matrix metalloproteinase processes These roles highlight the significance of VISTA/B7-H5 in immune regulation embryonic development and extracellular matrix dynamics VISTA/B7-H5 Protein Rat (HEK293 Fc) is the recombinant rat-derived VISTA/B7-H5 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-80141 Supplier Medchemexpress LLC Supplier No. HYP7354610UG
Add to Cart
Add to Cart
VISTA/B7-H5 protein an immunoregulatory receptor inhibits T-cell response and may regulate embryonic stem cell differentiation by inhibiting BMP4 signaling It also stimulates MMP14-mediated MMP2 activation suggesting a role in matrix metalloproteinase processes These roles highlight the significance of VISTA/B7-H5 in immune regulation embryonic development and extracellular matrix dynamics VISTA/B7-H5 Protein Rat (HEK293 Fc) is the recombinant rat-derived VISTA/B7-H5 protein expressed by HEK293 with C-hFc labeled tag