Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VISTA Detection Set (Risk Free), ProSci
SDP

Catalog No. 16595274 Shop All ProSci Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-274 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-274 Supplier ProSci LLC Supplier No. RF16070SET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Detection Sets

VISTA Monoclonal specifically detects VISTA in Human samples. It is validated for ELISA, Western Blot, ICC, IHC-P, Flow, IF

VISTA Antibody: VISTA/B7-H5/platelet receptor Gi24 is a single-pass type I membrane protein located at the cell surface. It is an immunoregulatory receptor which can inhibit T-cell response and may promote differentiation of embryonic stem cells, by inhibiting the BMP4 signaling pathway. The protein can be cleaved by MMP14, and stimulate MMP14-mediated MMP2 activation. ProSci's Risk-FreeTM antibodies are mouse monoclonal antibodies made to improve in vivo studies. Unlike antibodies developed using proteins made in yeast or bacteria, Risk-FreeTM antibodies are developed with antigens expressed in mammalian cell lines, giving the most native post-translational modifications to the protein. Validated for flow cytometry and ELISA Rigorously tested for the following applications: Immunoblot Immunohistochemistry Immunocytochemistry Immunofluorescence Multiple antibodies per target allowing the user to choose the best antibody for their application Available individually or as a set Risk-FreeTM means they are guaranteed to work for their approved applications

Specifications

Antigen VISTA
Applications ELISA, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence
Classification Monoclonal
Concentration Antibody 1 mg/mL
Conjugate Unconjugated
Formulation PBS containing 0.02% sodium azide.
Host Species Mouse
Purification Method Antibodies are supplied as protein A purified IgG1 and IgG2b.
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Antibodies can be stored at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Product Type Detection Sets
Form Liquid
Isotype IgG1, IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.