Learn More
Enzo Life Sciences VISTA monoclonal antibody (4C4) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS7000100
Encompass_Preferred
Clone: 4C4. Isotype: IgG1. Immunogen: Extracellular domain of human VISTA. Species reactivity: Human. Applications: Flow Cytometry, ICC, IF, IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-3239