Learn More
Hunter Lab Vista Transmission Spectrophotometer
SDP
Catalog No. NC1151313
Encompass
Vista spectrophotometer with the unique ability to capture both visible range transmission color and haze with a single measurement.
Catalog No. NC1151313 Supplier Hunter Lab Supplier No. VISTA
Add to Cart
Add to Cart
Request a Quote
Vista spectrophotometer with the unique ability to capture both visible range transmission color and haze with a single measurement.