Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies Pipette Tips: Low Retention
GREENER_CHOICESDP

Catalog No. 22146563 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Volume (Metric):
2 μL
25 μL
250 μL
1250 μL
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filtered Sterility Volume (Metric)
22-146-563 Filtered Sterile 2 μL
22-147-630 Non-filtered Non-sterile 1250 μL
22-146-560 Non-filtered Non-sterile 25 μL
22-147-631 Non-filtered Sterile 1250 μL
22-147-632 Filtered Sterile 1250 μL
22-146-561 Non-filtered Sterile 25 μL
22-146-564 Non-filtered Non-sterile 250 μL
22-146-565 Non-filtered Sterile 250 μL
22-146-562 Filtered Sterile 25 μL
22-146-566 Filtered Sterile 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 22-146-563 Supplier VistaLab™ Technologies Supplier No. 40701333LR
Add to Cart
Add to Cart

For use with Ovation and MLA Brand pipetters

  • Sterile, Low Retention tips packaged in VistaRak: 5 racks of 192 tips each
  • Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, etc.
  • Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
  • All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time

Specifications

Description Low Retention
Volume (Metric) 2 μL
Filtered Filtered
Sterility Sterile
Color Clear
Rack Color Blue
Autoclavable Not Autoclavable
Length (English) 1.7 in.
Thickness 5 in.
For Use With (Application) Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, TRIS, BSA and Tween, PCR or other biological applications
Sterilization Method e-beam
Format Rack
Tip Style Low Retention, Straight
Packaging VistaRak: 5 racks
Quantity 192 Tips/Rack
Product Type Pipette Tips
Color Code Blue
Disposable Single-use
For Use With (Equipment) Ovation
Graduated No
Material Polypropylene, Polyethylene (Filter)
Shape Cone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.