Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies MLA™ Brand Precision Pipetters
GSA_VAGREENER_CHOICESDP

Catalog No. 21324 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
25 μL
50 μL
100 μL
150 μL
200 μL
250 μL
300 μL
500 μL
1000 μL
Color:
Black
Blue
Brown
Gray
Green
Orange
Purple
Red
Silver
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
21-324 25 μL White
21-320 10 μL Orange
21-322 20 μL Black
21-326 50 μL Green
21-328 100 μL Blue
21-381-62 150 μL Silver
21-330 200 μL Red
21-332 250 μL Purple
21-334 300 μL Brown
21-336 500 μL Yellow
21-338 1000 μL Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 21-324 Supplier VistaLab™ Technologies Supplier No. 1023
Only null left
Add to Cart
Edge
Add to Cart

Ten fixed-volume models from 10 to 1000μL

  • 'To deliver' capability ensures volume accuracy over a wide range of viscosities
  • Ideal for repetitive delivery of reagents when high precision and accuracy are required
  • Sleeves are color coded for easy volume identification
  • Capacity engraved on bonnet

Order Info

Additional sizes available. Contact your Fisher Scientific Customer Service Representative for more information.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Accuracy ±3.00%
Autoclavable Not Autoclavable
Color White
Disposable Reusable
For Use With Micropipette Tip
Material Aluminum, Stainless Steel
Model F1
Number of Channels 1
Sterility Non-sterile
Type Precision Pipette
Volume (Metric) 25 μL
Includes Nozzle Insert, Calibration Key
Quantity 1 Ea.
Warranty Limited Lifetime
Length (English) 9 in.
Width (English) 2 in.
Height (English) 1.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.