Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies Reagent Reservoirs Accessory: Lid
GSA_VAGREENER_CHOICESDP

Catalog No. 21385110 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Capacity (Metric):
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
21-385-110 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-385-110 Supplier VistaLab™ Technologies Supplier No. 30543000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with 100mL reagent reservoirs

  • For 100mL reservoirs protects reagents from dust and minimize evaporation

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance RNase/DNase, Pyrogen-free
Color Clear
Capacity (Metric) 100 mL
Material Plastic
Sterility Sterile
Type Reservoir Lid
For Use With (Application) General Liquid Handling
Packaging Bulk
Quantity 100/Cs.
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.