Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Visual Management Face Mask with Shield Holder
SDP

Catalog No. 22000443
Change view
Click to view available options
Includes:
Keyhole slots for wall mount
Powerful Magnet Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-000-443 Powerful Magnet Mount
22-000-444 Keyhole slots for wall mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-000-443 Supplier Poltex Supplier No. FACESHLD2VMM
Add to Cart
Add to Cart

Visually manage and dispense PPE.

Durable and cleanable PETG construction

Specifications

Height (Metric) 393 mm
Width (Metric) 305 mm
Color Clear
Product Type Eyewear Dispenser
For Use With (Application) Face mask with shield
Includes Powerful Magnet Mount
Length (Metric) 183 mm
No. per Pack 1 Ea.
Material PETG

Consumables not included.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.