Learn More
Grainger Edwards Signaling Indicator Light, Amber, LED, 24V DC, 1 ea
Catalog No. 502791800
Encompass_Preferred
This amber LED beacon is a dual-mode signaling device designed for reliable visual indication in both indoor and outdoor environments. Engineered with a durable polycarbonate and ABS blend base and a shatter-resistant polycarbonate lens, this unit supports both steady-on and flashing operation modes, selectable via a dipswitch. It is rated for NEMA 4X enclosures, ensuring performance in demanding industrial settings.
  • Features selectable steady-on or flashing modes at 65 fpm
  • Compatible with panel or 1/2 in. NPT conduit mounting
  • Designed for indoor and outdoor applications
  • Rated for operation in temperatures ranging from -35°C to +66°C (-31°F to +150°F)
  • Includes shock, vibration, and corrosion resistance
  • Provides a projected LED lifespan of 148,000 hr.
  • UL/cUL listed for safety and compliance
Catalog No. 50-279-1800 Supplier Grainger Supplier No. 16G489
This amber LED beacon is a dual-mode signaling device designed for reliable visual indication in both indoor and outdoor environments. Engineered with a durable polycarbonate and ABS blend base and a shatter-resistant polycarbonate lens, this unit supports both steady-on and flashing operation modes, selectable via a dipswitch. It is rated for NEMA 4X enclosures, ensuring performance in demanding industrial settings.
  • Features selectable steady-on or flashing modes at 65 fpm
  • Compatible with panel or 1/2 in. NPT conduit mounting
  • Designed for indoor and outdoor applications
  • Rated for operation in temperatures ranging from -35°C to +66°C (-31°F to +150°F)
  • Includes shock, vibration, and corrosion resistance
  • Provides a projected LED lifespan of 148,000 hr.
  • UL/cUL listed for safety and compliance