The 105XBRi series is a heavy duty, multi-color LED visual signaling device designed for reliable performance in demanding environments. This indicator features a corrosion-resistant NEMA 4X enclosure and a shatter-resistant polycarbonate double fresnel lens, making it suitable for both indoor and outdoor marine applications. Utilizing XTRA-SAFE™ technology, the unit provides distinct status indications, including support for those who are color blind, and offers flexible operational modes via dipswitch settings.
- Corrosion-resistant type 4X enclosure suitable for marine use
- Shatter-resistant polycarbonate double fresnel lens
- Operational in temperatures from -35°C to 66°C (-31°F to 150°F)
- Supports steady or flashing light modes
- Designed for panel, conduit, or wall mounting
- Integrated LED technology with a 148,000 hr projected lifespan
- PLC compatible with