Learn More
Grainger Edwards Signaling LED Multi-Status Indicator, NEMA 4X, 24V DC, 1 ea
Catalog No. 502791928
Encompass_Preferred
This heavy duty LED multi-status indicator provides reliable visual signaling with a corrosion-resistant NEMA 4X enclosure. Designed for versatility, this beacon features an adaptive design with dip switch settings that allow for standalone operation or control via an external PLC, PAC, or control relay. The device is constructed with a durable, glass-reinforced thermoplastic polyester resin base and a shatter-resistant polycarbonate lens, making it suitable for demanding industrial and marine environments.
  • Features patented XTRA-SAFE™ technology using color and flash-rate combinations to assist those who are color-blind
  • Provides multiple mounting options including 3/4 in. pipe, outlet box, or wall bracket configurations
  • Rated for operation in temperature ranges from -35°C to 66°C
  • Offers a projected LED service life of 148,000 hours
  • Includes UL and cUL listings for marine use a
Catalog No. 50-279-1928 Supplier Grainger Supplier No. 16G485
This heavy duty LED multi-status indicator provides reliable visual signaling with a corrosion-resistant NEMA 4X enclosure. Designed for versatility, this beacon features an adaptive design with dip switch settings that allow for standalone operation or control via an external PLC, PAC, or control relay. The device is constructed with a durable, glass-reinforced thermoplastic polyester resin base and a shatter-resistant polycarbonate lens, making it suitable for demanding industrial and marine environments.
  • Features patented XTRA-SAFE™ technology using color and flash-rate combinations to assist those who are color-blind
  • Provides multiple mounting options including 3/4 in. pipe, outlet box, or wall bracket configurations
  • Rated for operation in temperature ranges from -35°C to 66°C
  • Offers a projected LED service life of 148,000 hours
  • Includes UL and cUL listings for marine use a