The 125XBRi Series LED Multi-Status Indicator is a versatile, multi-color visual signaling device designed for indoor and outdoor monitoring or alarm systems. This unit provides customizable alert patterns to signal hazardous conditions and is built with a rugged, corrosion-resistant housing suitable for demanding industrial environments.
- Features a NEMA 4X enclosure constructed from 33% glass-filled nylon, providing chemical, heat, and shatter resistance.
- Offers flexible installation with panel or conduit mounting options, including 1/2 in. FNPT and 3/4 in. MNPT threads.
- Provides fully customizable activation of colors and flash rates when controlled by a PLC or external control relay.
- Equipped with an energy-efficient LED source offering an operating life of 148,000 hr.
- Designed for operation at 24 VDC with an operating current of 0.150 A.
- Functions within a broad temperature range