Learn More
Grainger Edwards Signaling LED Beacon, Multi-Status, 24 VDC, 1 ea
Catalog No. 502792383
Encompass_Preferred
The 125XBRi Series multi-status indicator is a versatile 24 VDC beacon designed for reliable signaling in demanding environments. This indicator utilizes patented XTRA-SAFE technology, which combines specific color and flash-rate patterns to provide clear status information, including compatibility for those who are color blind. Its robust construction ensures longevity and performance in industrial applications.
  • Features XTRA-SAFE technology with dipswitch-enabled flash modes
  • Includes a 33% glass-filled nylon base for high heat and chemical resistance
  • Utilizes a shatter-resistant polycarbonate lens
  • Offers an enclosure rating of NEMA 4X for corrosion resistance
  • Designed for panel or conduit mounting with 1/2 in. internal or 3/4 in. external hubs
  • Operates in temperatures from -35°C to 66°C
  • Provides a long operational life of 148,000 hours
Catalog No. 50-279-2383 Supplier Grainger Supplier No. 16G819
The 125XBRi Series multi-status indicator is a versatile 24 VDC beacon designed for reliable signaling in demanding environments. This indicator utilizes patented XTRA-SAFE technology, which combines specific color and flash-rate patterns to provide clear status information, including compatibility for those who are color blind. Its robust construction ensures longevity and performance in industrial applications.
  • Features XTRA-SAFE technology with dipswitch-enabled flash modes
  • Includes a 33% glass-filled nylon base for high heat and chemical resistance
  • Utilizes a shatter-resistant polycarbonate lens
  • Offers an enclosure rating of NEMA 4X for corrosion resistance
  • Designed for panel or conduit mounting with 1/2 in. internal or 3/4 in. external hubs
  • Operates in temperatures from -35°C to 66°C
  • Provides a long operational life of 148,000 hours