The 105XBRi Series XTRA-BRITE™ LED multi-status indicator is a heavy duty visual signaling device designed for reliable performance in demanding environments. This beacon features advanced LED technology to provide clear, multi-color status indication suitable for various indoor and outdoor applications.
- Equipped with XTRA-SAFE™ technology for color-blind accessibility
- Operates at 120V AC with 0.1 A current draw
- Constructed with a glass reinforced thermoplastic polyester resin housing and polycarbonate lens
- Offers NEMA 4X protection and immunity to shock and vibration
- Supports multiple mounting options including 3/4 in. pipe, outlet box, or wall mount
- Features steady light mode with selectable flash rates of 65, 120, or 240 per minute
- Rated for operation in temperatures ranging from -31°F to 150°F
- Suitable for Class I, Division 2, Class II, Divisio