Learn More
Grainger Edwards Signaling Indicator Light, Green, LED, 24V DC, 1 ea
Catalog No. 502791671
Encompass_Preferred
The 48XBRM Series LED dual-mode signaling beacon provides reliable visual indication in demanding environments. Designed for both indoor and outdoor applications, this beacon features an impact- and vibration-resistant construction, making it suitable for high-noise areas or settings where long service life is required. The unit ships in steady-on mode and can be configured for a 65 FPM flashing pattern via an internal dipswitch. It is engineered for use with 24 V DC power sources and utilizes a durable polycarbonate lens.
  • Features dual-mode operation (steady-on or 65 FPM flashing)
  • Constructed for high-vibration and high-noise environments
  • NEMA 4X rated for indoor or outdoor use
  • Long lamp life of 148,000 hrs.
  • Designed for 24 V DC operation with 0.215 A current draw
  • Includes a 1/2 in. conduit hole for mounting
  • Constructed with impact-, shatter-, and corrosion-resistant materia
Catalog No. 50-279-1671 Supplier Grainger Supplier No. 16G493
The 48XBRM Series LED dual-mode signaling beacon provides reliable visual indication in demanding environments. Designed for both indoor and outdoor applications, this beacon features an impact- and vibration-resistant construction, making it suitable for high-noise areas or settings where long service life is required. The unit ships in steady-on mode and can be configured for a 65 FPM flashing pattern via an internal dipswitch. It is engineered for use with 24 V DC power sources and utilizes a durable polycarbonate lens.
  • Features dual-mode operation (steady-on or 65 FPM flashing)
  • Constructed for high-vibration and high-noise environments
  • NEMA 4X rated for indoor or outdoor use
  • Long lamp life of 148,000 hrs.
  • Designed for 24 V DC operation with 0.215 A current draw
  • Includes a 1/2 in. conduit hole for mounting
  • Constructed with impact-, shatter-, and corrosion-resistant materia