Learn More
Medchemexpress LLC VIT-2763 | 2095668-10-1 | 99.9% | 408.43 | 50 MG
SDP
Catalog No. 5000437853
Encompass_Preferred
VIT-2763 is an oral ferroportin inhibitor designed to inhibit hepcidin binding to ferroportin and block iron efflux. It holds potential for treating β-thalassemia by addressing iron regulation. This compound has demonstrated significant biological activity both in vitro and in vivo.
  • Reduces fluorescence polarization signal by displacing TMR-hepcidin from ferroportin.
  • Induces BLA reporter gene activity, increasing intracellular iron concentrations by blocking iron export.
  • Triggers ubiquitination and degradation of ferroportin.
  • Corrects anemia and improves red blood cell parameters in relevant animal models.
  • Decreases the percentage of ROS-positive red blood cells and extends their lifespan.
  • Significantly decreases serum iron levels in animal studies.
  • Currently undergoing Phase 2 clinical trials for Beta-Thalassemia and Sickle Cell Disease.
Catalog No. 50-004-37853 Supplier Medchemexpress LLC Supplier No. HY11222050MG
VIT-2763 is an oral ferroportin inhibitor designed to inhibit hepcidin binding to ferroportin and block iron efflux. It holds potential for treating β-thalassemia by addressing iron regulation. This compound has demonstrated significant biological activity both in vitro and in vivo.
  • Reduces fluorescence polarization signal by displacing TMR-hepcidin from ferroportin.
  • Induces BLA reporter gene activity, increasing intracellular iron concentrations by blocking iron export.
  • Triggers ubiquitination and degradation of ferroportin.
  • Corrects anemia and improves red blood cell parameters in relevant animal models.
  • Decreases the percentage of ROS-positive red blood cells and extends their lifespan.
  • Significantly decreases serum iron levels in animal studies.
  • Currently undergoing Phase 2 clinical trials for Beta-Thalassemia and Sickle Cell Disease.