Learn More
Medchemexpress LLC HY-112220 5mg , VIT-2763 CAS:2095668-10-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1122205MG
Encompass_Preferred
HY-112220 5mg VIT-2763 CAS: 2095668-10-1 VIT-2763, an oral ferroportin inhibitor, inhibits hepcidin binding to ferroportin and blocks iron efflux. VIT-2763 has the potential in the treatment of β-thalassemia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9630