Learn More
NEXPRING HEALTH Vit Kit-Freeze
Catalog No. NC1256792
Encompass
Vit Kit Freeze is a flexible, cost-effective system for use in the vitrification of oocytes (MII), pronuclear (PN) zygotes, cleavage stage embryos, and blastocyst stage embryos. With survival rates ranging between 90-99%, and featuring low-volume protocols, Vit Kit Freeze can support your laboratory by reducing the cost of vitrification and ensuring consistent, high-survival rates.
Catalog No. NC1256792 Supplier NEXPRING HEALTH Supplier No. 90133SO
Vit Kit Freeze is a flexible, cost-effective system for use in the vitrification of oocytes (MII), pronuclear (PN) zygotes, cleavage stage embryos, and blastocyst stage embryos. With survival rates ranging between 90-99%, and featuring low-volume protocols, Vit Kit Freeze can support your laboratory by reducing the cost of vitrification and ensuring consistent, high-survival rates.