Learn More
NEXPRING HEALTH Vit Kit-Thaw
Catalog No. NC1256793
Encompass
Vit Kit Thaw is a flexible, cost-effective system for use in the thawing of oocytes (MII), pronuclear (PN) zygotes, cleavage stage embryos, and blastocyst stage embryos. With survival rates ranging from 90-99%, and featuring low-volume protocols, Vit Kit Thaw can support your laboratory by reducing the cost of vitrification and ensuring consistent, high-survival rates.
Catalog No. NC1256793 Supplier NEXPRING HEALTH Supplier No. 90137SO
Vit Kit Thaw is a flexible, cost-effective system for use in the thawing of oocytes (MII), pronuclear (PN) zygotes, cleavage stage embryos, and blastocyst stage embryos. With survival rates ranging from 90-99%, and featuring low-volume protocols, Vit Kit Thaw can support your laboratory by reducing the cost of vitrification and ensuring consistent, high-survival rates.