Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Vitachrome™ Live Amoeba proteus
SDPSCI_ED

Catalog No. S42848 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Sufficient For:
Class of 30 students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
S42848 Class of 30 students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S42848 Supplier Carolina™ Supplier No. 131308

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Prestained living organisms used for teaching and photomicrography.

  • Prestained for classroom use
  • Sharp, clear staining of cultures provides means for the presentation of internal structure
  • Contains enough material for class of 30 students

Specifications

For Use With (Application) Teaching and photomicrography
Quantity 1 Ea.
Scientific Name Amoeba proteus
Includes Dropping pipet
Sufficient For Class of 30 students
Product Type Protozoa Mixture
Description Prestained
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.