Learn More
Selleck Chemical LLC Vitamin A-25mg
SDP
Supplier:  Selleck Chemical LLC S559225MG
Encompass_Preferred
Vitamin A (Retinol) is a naturally occuring fat-soluble vitamin that is important for normal vision, the immune system, and reproduction. It also plays roles in normal functioning of heart, lungs, kidneys, and other organs.
Catalog No. 50-238-9690