Learn More
Medchemexpress LLC Vitamin A | 68-26-8 | 98.3% | 100 MG
SDP
Catalog No. 5000412068
Encompass_Preferred
Vitamin A (Vitamin A1) is an endogenous metabolite for research use only. This compound is stable under recommended storage conditions, which include protection from light and storage under nitrogen. It is important to note that the compound is unstable in solutions, and freshly prepared solutions are recommended for use.
  • Suitable for laboratory research
  • Recommended for experienced personnel
  • Stable under specific storage conditions
  • Requires freshly prepared solutions for use
Catalog No. 50-004-12068 Supplier Medchemexpress LLC Supplier No. HYB1342100MG
Vitamin A (Vitamin A1) is an endogenous metabolite for research use only. This compound is stable under recommended storage conditions, which include protection from light and storage under nitrogen. It is important to note that the compound is unstable in solutions, and freshly prepared solutions are recommended for use.
  • Suitable for laboratory research
  • Recommended for experienced personnel
  • Stable under specific storage conditions
  • Requires freshly prepared solutions for use