Learn More
Enzo Life Sciences Vitamin A acetate (5g). CAS: 127-47-9
SDP
Catalog No. 502009295
Encompass_Preferred
Alternative name: Retinyl acetate. Purity: ≥95% (HPLC). Solubility: Soluble in ethanol, chloroform, and ether. Insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9295 Supplier Enzo Life Sciences Supplier No. ALX4600390005
Alternative name: Retinyl acetate. Purity: ≥95% (HPLC). Solubility: Soluble in ethanol, chloroform, and ether. Insoluble in water. Long Term Storage: -20°C.