Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin A Palmitate, 1.70 MIU/g, USP, 1.62 MIU/g, Spectrum™ Chemical
SDP

Catalog No. 18607713 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-713 100 mL Amber Glass Bottle
18-607-712 25 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-607-713 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. V1159100ML
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

V1159, 79-81-2, C36H60O2

Spectrum™ Chemical Vitamin A Palmitate, 1.70 MIU/g, USP is also known as retinyl acetate or retinol acetate and is often used to fortify foods with vitamin A. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 79-81-2
Molecular Formula C36H60O2
Molecular Weight (g/mol) 524.87
InChI Key VYGQUTWHTHXGQB-FFHKNEKCSA-N
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C

Specifications

CAS Max % 1
Color Yellow to Orange
Packaging Amber Glass Bottle
Quantity 100 mL
Percent Purity 1.62 MIU/g
Grade USP
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Vitamin A Palmitate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.