Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin A palmitate, MP Biomedicals™

Catalog No. ICN10326380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10326380 100 g
ICN10326305 5 g
ICN10326325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10326380 Supplier MP Biomedicals, Inc Supplier No. 0210326380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79-81-2
Molecular Formula C36H60O2
Molecular Weight (g/mol) 524.874
InChI Key VYGQUTWHTHXGQB-FFHKNEKCSA-N
Synonym retinyl palmitate, vitamin a palmitate, retinol palmitate, retinol, hexadecanoate, arovit, all-trans-retinyl palmitate, optovit-a, retinyl hexadecanoate, aquapalm, dispatabs tabs
PubChem CID 5280531
ChEBI CHEBI:17616
IUPAC Name [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC=C(C)C=CC=C(C)C=CC1=C(CCCC1(C)C)C

Specifications

Color Yellow to Orange
Quantity 100 g
Formula Weight 524.9
Physical Form Powder
Chemical Name or Material Vitamin A palmitate
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.