Learn More
Medchemexpress LLC Vitamin A Propionate | 7069-42-3 | 98.91% | 342.51 | 500 MG
SDP
Catalog No. 5000451808
Encompass_Preferred
Vitamin A Propionate is an ester compound of vitamin A. In nutritional studies of foals, different doses have different effects on growth, serum biochemistry, and hematological indicators. Too high or too low doses will produce adverse effects and are related to the vitamin A concentration in plasma, liver, and kidneys.
  • For research use only.
  • Not sold to patients.
Catalog No. 50-004-51808 Supplier Medchemexpress LLC Supplier No. HYW011180500MG
Vitamin A Propionate is an ester compound of vitamin A. In nutritional studies of foals, different doses have different effects on growth, serum biochemistry, and hematological indicators. Too high or too low doses will produce adverse effects and are related to the vitamin A concentration in plasma, liver, and kidneys.
  • For research use only.
  • Not sold to patients.