Learn More
Biomatik Corporation Vitamin B1 (VB1) CLIA Kit
Supplier:  Biomatik Corporation EKU09502
Encompass_Preferred
Vitamin B1 (VB1) CLIA Kit for detecting VB1 in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 1.17-300ng/ml. Sensitivity: 0.41ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Target Synonyms: Thiamine; Thiamin; Aneurine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8509