Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin B12, 0.1% in Mannitol, ∽96%, MP Biomedicals™

Catalog No. ICN10327810 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10327810 10 g
ICN10327825 25 g
ICN10327880 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10327810 Supplier MP Biomedicals, Inc Supplier No. 0210327810
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68-19-9
Molecular Formula C63H88CoN14O14P
Molecular Weight (g/mol) 1355.39
MDL Number MFCD00151092
InChI Key YUWVGNQGDAPKCX-BVWPOUIRNA-L
Synonym vitamin b12
PubChem CID 129893524
IUPAC Name (10S,12R,13R,24S,35R,41R)-30,35,40-tris(2-carbamoylethyl)-24,36,41-tris(carbamoylmethyl)-1-cyano-46-hydroxy-12-(hydroxymethyl)-5,6,17,23,28,31,31,36,38,41,42-undecamethyl-15,20-dioxo-11,14,16-trioxa-2λ⁵,9,19,26λ⁵,43,44λ⁵,45λ⁵-heptaaza-15λ⁵-phospha-1-cobaltadodecacyclo[27.14.1.1¹,³⁴.1²,⁹.1¹⁰,¹³.0¹,²⁶.0³,⁸.0²³,²⁷.0²⁵,⁴².0³²,⁴⁴.0³⁹,⁴³.0³⁷,⁴⁵]heptatetraconta-2(47),3,5,7,26,28,32(44),33,37(45),38-decaene-1,2,26,44,45-pentakis(ylium)-15-olate
SMILES [C@@H]1(C2=CC3=[N+]4C(C(C3(C)C)CCC(N)=O)=C(C3=[N+]5C6C7(N8C(C([C@@]7(C)CC(N)=O)CCC(N)=O)=C(C(=[N+]2[Co+]458([N+]2=CN(C4=CC(=C(C)C=C24)C)[C@@]2([H])O[C@@H]([C@]([H])(OP([O-])(=O)OC(C)CNC(=O)CCC3(C)[C@@H]6CC(N)=O)C2O)CO)C#N)C1(C)CC(N)=O)C)C)C)CCC(N)=O

Specifications

Color White
Quantity 10 g
Formula Weight 1355.42
Physical Form Powder
Chemical Name or Material Vitamin B12
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.