Learn More
Apexbio Technology LLC Vitamin B12 68-19-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062597
Encompass_Preferred
Vitamin B12 (CAS 68-19-9) is a cobalt-containing water-soluble vitamin involved in hematopoiesis and neuronal function It acts primarily as a cofactor facilitating methyl-transfer reactions notably as methylcobalamin in the synthesis pathway of methionine consequently regulating methylation of DNA RNA and proteins Additionally Vitamin B12 modulates cellular redox homeostasis offering protective effects against oxidative cellular damage Current research engages Vitamin B12 in studies on neurodegenerative disease models immunological inflammation pathways and cellular injury mechanisms related to oxidative stress
Catalog No. 50-000-62597 Supplier Apexbio Technology LLC Supplier No. B20635.1
Vitamin B12 (CAS 68-19-9) is a cobalt-containing water-soluble vitamin involved in hematopoiesis and neuronal function It acts primarily as a cofactor facilitating methyl-transfer reactions notably as methylcobalamin in the synthesis pathway of methionine consequently regulating methylation of DNA RNA and proteins Additionally Vitamin B12 modulates cellular redox homeostasis offering protective effects against oxidative cellular damage Current research engages Vitamin B12 in studies on neurodegenerative disease models immunological inflammation pathways and cellular injury mechanisms related to oxidative stress