Learn More
Enzo Life Sciences Vitamin B12 (5g). CAS: 68-19-9
SDP
Supplier:  Enzo Life Sciences ALX460024G005
Encompass_Preferred
Alternative name: Cyanocobalamin. Purity: ≥96% (HPLC). Solubility: Soluble in water (12.5mg/ml) or ethanol. Long Term Storage: +4°C.
Catalog No. 50-200-9289