Learn More
MOLECULAR DEPOT LLC Vitamin B12 BSA - 0.5 mg
Catalog No. 5000013484
Encompass_Preferred
Vitamin B12 BSA Conjugate (0 5 mg) consists of vitamin B12 conjugated to Bovine Serum Albumin (BSA) through a flexible PEG linker This product has been used as a molecular tool in the study of the metabolism of vitamin B12 as well as the development of vitamin B12 detection methods It supplied as a solution For in vitro research use only
Catalog No. 50-000-13484 Supplier MOLECULAR DEPOT LLC Supplier No. B2010606
Vitamin B12 BSA Conjugate (0 5 mg) consists of vitamin B12 conjugated to Bovine Serum Albumin (BSA) through a flexible PEG linker This product has been used as a molecular tool in the study of the metabolism of vitamin B12 as well as the development of vitamin B12 detection methods It supplied as a solution For in vitro research use only