Learn More
MOLECULAR DEPOT LLC Vitamin B12 Diluent - 50 mL
Catalog No. 5000013426
Encompass_Preferred
Vitamin B12 Buffered Diluent is a liquid-stable buffered solution containing undetectable levels of vitamin B12 (less than 75 pg/mL) This product has been used in the research and development of vitamin B12 high and low working solutions This product is for research use only
Catalog No. 50-000-13426 Supplier MOLECULAR DEPOT LLC Supplier No. B2010583
Vitamin B12 Buffered Diluent is a liquid-stable buffered solution containing undetectable levels of vitamin B12 (less than 75 pg/mL) This product has been used in the research and development of vitamin B12 high and low working solutions This product is for research use only