Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin B12, MP Biomedicals™

Catalog No. MP021032715 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021032715 500 mg
MP021032712 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021032715 Supplier MP Biomedicals, Inc Supplier No. 02103271.5
Add to Cart
Add to Cart

Plays a role in myelin synthesis and is used an antipernicious anemia factor. Vitamin B12, MP Biomedicals (cobalamin) is a group of chemically-related cobalt-containing molecules involved in cell processes such as DNA synthesis, fatty acid synthesis, ener

The physiologically active forms of vitamin B12 include methylcobalamin and adenosylcobalamin, whereas hydroxocobalamin (vitamin B12a, OHCbl) and cyanocobalamin (CNCbl) are storage and delivery forms.

Biosynthesis of the basic structure of the vitamin is accomplished only by bacteria (which usually produce hydroxocobalamin). Bacteria-derived hydroxocobalamin (OHCbl) and CNCbl are converted in humans to useful coenzyme forms to support metabolic processes, such as mitochondrial methylmalonylcoenzyme A mutase conversion of methylmalonic acid (MMA) to succinate to link lipid and carbohydrate metabolism, and activation of methionine synthase, the rate limiting step in the synthesis of methionine.

  • Vitamin B12 plays a role in the synthesis of myelin
  • It is a cofactor for two enzymes, methionine synthase and L-methylmalonyl-CoA mutase
  • The compound is also used as an antipernicious anemia factor
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-19-9
Molecular Formula C63H88CoN14O14P
Molecular Weight (g/mol) 1355.39
MDL Number MFCD00151092
InChI Key YUWVGNQGDAPKCX-BVWPOUIRNA-L
Synonym vitamin b12
PubChem CID 129893524
IUPAC Name (10S,12R,13R,24S,35R,41R)-30,35,40-tris(2-carbamoylethyl)-24,36,41-tris(carbamoylmethyl)-1-cyano-46-hydroxy-12-(hydroxymethyl)-5,6,17,23,28,31,31,36,38,41,42-undecamethyl-15,20-dioxo-11,14,16-trioxa-2λ⁵,9,19,26λ⁵,43,44λ⁵,45λ⁵-heptaaza-15λ⁵-phospha-1-cobaltadodecacyclo[27.14.1.1¹,³⁴.1²,⁹.1¹⁰,¹³.0¹,²⁶.0³,⁸.0²³,²⁷.0²⁵,⁴².0³²,⁴⁴.0³⁹,⁴³.0³⁷,⁴⁵]heptatetraconta-2(47),3,5,7,26,28,32(44),33,37(45),38-decaene-1,2,26,44,45-pentakis(ylium)-15-olate
SMILES [C@@H]1(C2=CC3=[N+]4C(C(C3(C)C)CCC(N)=O)=C(C3=[N+]5C6C7(N8C(C([C@@]7(C)CC(N)=O)CCC(N)=O)=C(C(=[N+]2[Co+]458([N+]2=CN(C4=CC(=C(C)C=C24)C)[C@@]2([H])O[C@@H]([C@]([H])(OP([O-])(=O)OC(C)CNC(=O)CCC3(C)[C@@H]6CC(N)=O)C2O)CO)C#N)C1(C)CC(N)=O)C)C)C)CCC(N)=O

Specifications

Melting Point >300°C (literature)
Quantity 500 mg
Solubility Information Soluble in alcohol; slightly soluble in H2O.
Formula Weight 1355.42
Percent Purity ≥96%
Physical Form Powder
Recommended Storage 4°C, desiccate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.