Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin B12a hydrochloride, ≥95%, MilliporeSigma™ Supelco™

Catalog No. 111014800 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4800 100 mg
11-101-4799 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4800 Supplier MilliporeSigma Supelco Supplier No. H7126100MG
Add to Cart
Add to Cart

Vitamin B12a is an organic derivative, used as a powerful antidote for cyanide poisoning. It is a storage and delivery form of vitamin B12 and its mode of action involves the formation of non-toxic cyanocobalamin.

Vitamins, Nutraceuticals and Natural Products
TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 59461-30-2
Physical Form Neat
Linear Formula C62H89 CoN13O15 P · HCl
MDL Number MFCD00167463
Synonym Hydroxocobalamin hydrochloride
Type Analytical Standard
Quantity 100 mg
Formula Weight 1382.82
Percent Purity ≥95%
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.