Learn More
Cayman Chemical Vitamin B12, Purity: ≥95%, CAS: 68-19-9, 500 mg
SDP
Catalog No. 502966142
Encompass_Preferred
A corrinoid vitamin with roles in brain and nervous system functioning and blood formation through regulation of DNA synthesis, cellular metabolism, fatty acid metabolism, and amino acid metabolism; serves as a cofactor in isomerase-, methyltransferase-, and dehalogenase-catalyzed reactions, including the conversion of methylmalonyl-CoA to succinyl-CoA and the transfer of a methyl group from 5-methyltetrahydrofolate to homocysteine
Catalog No. 50-296-6142 Supplier Cayman Chemical Supplier No. 18425500 MG
A corrinoid vitamin with roles in brain and nervous system functioning and blood formation through regulation of DNA synthesis, cellular metabolism, fatty acid metabolism, and amino acid metabolism; serves as a cofactor in isomerase-, methyltransferase-, and dehalogenase-catalyzed reactions, including the conversion of methylmalonyl-CoA to succinyl-CoA and the transfer of a methyl group from 5-methyltetrahydrofolate to homocysteine