Learn More
Cambridge Isotope Laboratories DEUTERIUM CHLORIDE (D, 99%), 5 L, 7698-05-7
Supplier:  Cambridge Isotope Laboratories DLM4585
Encompass
DEUTERIUM CHLORIDE (D, 99%), 5 L, 7698-05-7. This item is shipped in a Code K-460 stainless steel cylinder. The fee for the cylinder will need added to the order as a separate charge per each. Please include part# NC3244894 to the order.
Catalog No. 50-163-1097