Learn More
Medchemexpress LLC Vitamin B5-d4 calcium | 20938-62-9 | 99.17% | C18H24D8CaN2O10 | 1 MG
SDP
Catalog No. 5000354512
Encompass_Preferred
Vitamin B5-d4 calcium is a deuterium labeled Vitamin B5 calcium, used for research purposes. It can serve as a tracer or an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Stable heavy isotopes, such as hydrogen and carbon, are incorporated into drug molecules primarily for quantitation during drug development, and deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled vitamin B5 calcium
  • Functions as a tracer in research applications
  • Acts as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation during drug development
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-54512 Supplier Medchemexpress LLC Supplier No. HY144243S1MG
Vitamin B5-d4 calcium is a deuterium labeled Vitamin B5 calcium, used for research purposes. It can serve as a tracer or an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Stable heavy isotopes, such as hydrogen and carbon, are incorporated into drug molecules primarily for quantitation during drug development, and deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled vitamin B5 calcium
  • Functions as a tracer in research applications
  • Acts as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation during drug development
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs