Learn More
Sigma Aldrich Fine Chemicals Biosciences Vitamin B5 di beta alanine5MG
Catalog No. 502905877
Encompass
Vitamin B5 (di--alanine-13C6 15N2) calcium salt is also known as calcium pantothenate-[13C6 15N2]. Vitamin B5 (a B vitamin) also known as pantothenic acid is a water-soluble vitamin found in animal and plant products. Vitamin B5 deficiency causes dyslipidemia (abnormal levels of lipids in the blood) and dermatitis.
Catalog No. 50-290-5877 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 7058375MG
Vitamin B5 (di--alanine-13C6 15N2) calcium salt is also known as calcium pantothenate-[13C6 15N2]. Vitamin B5 (a B vitamin) also known as pantothenic acid is a water-soluble vitamin found in animal and plant products. Vitamin B5 deficiency causes dyslipidemia (abnormal levels of lipids in the blood) and dermatitis.