Learn More
TARGETMOL CHEMICALS INC Vitamin B6 5MG
SDP
Catalog No. 502877125
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Vitamin B6 is a component of some coenzymes in human body, which is involved in a variety of metabolic reactions, especially related to amino acid metabolism.Clinical application of vitamin B6 in the prevention and treatment of pregnancy and radiation sickness vomiting. Purity 100%
Catalog No. 50-287-7125 Supplier TARGETMOL CHEMICALS INC Supplier No. T3O27845MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Vitamin B6 is a component of some coenzymes in human body, which is involved in a variety of metabolic reactions, especially related to amino acid metabolism.Clinical application of vitamin B6 in the prevention and treatment of pregnancy and radiation sickness vomiting. Purity 100%